Dipropyleneglycol dimethyl ether (mixture of isomers) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Dipropylene glycol dimethyl ether
M-Clarity™ quality level = MQ200
8.43825.0250
8.43825.1000
8.43825.0250
8.43825.1000
EM8.43825.1000CS
780
USD
EM8.43825.1000
EM8.43825.0250
CA8.43825.0250
CA8.43825.1000
Dipropyleneglycol dimethyl ether (mixture of isomers) for synthesis, Sigma-Aldrich®
Dipropyleneglycol dimethyl ether (mixture of isomers)
Formula:
C₈H₁₈O₃ |
MDL Number:
MFCD00210047 CAS Number: 111109-77-4 |
Specification Test Results
Solubility | 526 g/l |
Melting Point | -80 °C |
Molar Mass | 162.23 g/mol |
Boiling Point | 175 °C (1013 hPa) |
Vapor Pressure | 0.8 hPa (20 °C) |
Flash Point | 65 °C |
Explosion Limit | 0.77 - 5.57 %(V) |
Density | 0.904 g/cm3 |
pH | (100 g/l, H2O, 20 °C) neutral |
Ignition Point | 165 °C DIN 51794 |
Viscosity kinematic | 1.12 mm2/s (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...