Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3,5-Triiodobenzoic acid for synthesis

Supplier: MilliporeSigma

Warning
Synonyms: TIBA

8.41852.0005 8.41852.0005
EM8.41852.0005CS 33088 USD InStock
EM8.41852.0005 CA8.41852.0005
2,3,5-Triiodobenzoic acid for synthesis
2,3,5-Triiodobenzoic acid


Formula: C₇H₃I₃O₂
MW: 499.81 g/mol
Boiling Pt: 416 °C (1013 hPa)
Melting Pt: 225,9 °C
Storage Temperature: Freezer
MDL Number: MFCD00002420
CAS Number: 88-82-4
EINECS: 201-859-6
Order Now
Specification Test Results
Melting Point 226 °C
Molar Mass 499.81 g/mol


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty