Trimethyloxonium tetrafluoroborate ≥95.0% (by acidimetry) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.41638.0010 8.41638.0010
EM8.41638.0010CS 3996 USD
CA8.41638.0010 EM8.41638.0010
Trimethyloxonium tetrafluoroborate ≥95.0% (by acidimetry) for synthesis, Sigma-Aldrich®
Trimethyloxonium tetrafluoroborate
Formula: C₃H₉BF₄O
Storage Temperature: Freezer
MDL Number: MFCD00011798
CAS Number: 420-37-1
EINECS: 206-994-4
UN: 3131
ADR: 8,II

Order Now

Specification Test Results

Assay (acidimetric) ≥ 95.0 %
Identity (IR) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR