1,1'-[Oxybis(ethyleneoxy)]diethylene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 1,1'-(Oxybis(ethyleneoxy))dietheneDiethylene glycol divinyl ether(2-(2-Ethenyloxyethoxy)ethoxy)ethene1,1'-[Oxybis(ethyleneoxy)]diethene1,1'-(Oxybis(ethyleneoxy))diethylene

M-Clarity™ quality level = MQ200

8.40144.0050 8.40144.0050
EM8.40144.0050CS 18272 USD
CA8.40144.0050 EM8.40144.0050
1,1'-[Oxybis(ethyleneoxy)]diethylene for synthesis, Sigma-Aldrich®
1,1'-[Oxybis(ethyleneoxy)]diethylene
Formula: H₂CCHO(CH₂)₂O(CH₂)₂OCHCH₂
MW: 158.2 g/mol
Density: 0.97 g/cm³ (20 °C)
MDL Number: MFCD00054610
CAS Number: 764-99-8
EINECS: 212-133-3

Order Now

Specification Test Results

Molar Mass 158.20 g/mol
Density 0.97 g/cm3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR