Piperonyl butoxide for synthesis, Sigma-Aldrich®
|
Warning
|
Synonyms:
5-((2-(2-Butoxyethoxy)ethoxy)methyl)-6-propylbenzo[d][1,3]dioxole
, 5-[2-(2-Butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole
, NSC 8401
M-Clarity™ quality level = MQ200
8.40080.0025
8.40080.0025
EM8.40080.0025CS
2064.97
USD
EM8.40080.0025
CA8.40080.0025
Piperonyl butoxide for synthesis, Sigma-Aldrich®
Piperonyl butoxide
Formula:
C₁₉H₃₀O₅ MW: 338.44 g/mol |
MDL Number:
MFCD00005842 CAS Number: 51-03-6 EINECS: 200-076-7 UN: 3082 ADR: 9,III |
Specification Test Results
Solubility | 0.01 g/l (25 °C) |
Molar Mass | 338.44 g/mol |
Boiling Point | 180 °C (1.3 hPa) |
Vapor Pressure | 0.02 hPa (60 °C) |
Flash Point | 212 °C |
Density | 1.06 g/cm3 (20 °C) |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous Hazardous Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...