Octadecyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
ODTS
, Trichloro(stearyl)silane
, Trichlorooctadecylsilane
, Trichloro(octadecyl)silane
, Octadecyltrichlorosilane
M-Clarity™ quality level = MQ200
8.22170.0100
8.22170.0100
EM8.22170.0100CS
2662
USD
EM8.22170.0100
CA8.22170.0100
Octadecyltrichlorosilane for synthesis, Sigma-Aldrich®
Octadecyltrichlorosilane
Formula:
C₁₈H₃₇Cl₃Si MW: 387.94 g/mol Boiling Pt: 361 °C (1013 hPa) Melting Pt: 22 °C Density: 0.98 g/cm³ (20 °C) Flash Pt: 89 °C |
MDL Number:
MFCD00000484 CAS Number: 112-04-9 EINECS: 203-930-7 UN: 1800 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Molar Mass | 387.94 g/mol |
Boiling Point | 223 °C (13 hPa) |
Flash Point | 89 °C |
Density | 1228 g/mol |
pH | (H2O, 20 °C) not applicable |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...