Lithium hydride for synthesis, Sigma-Aldrich®
|
Danger
|
M-Clarity™ quality level = MQ200
8.20753.0050
8.20753.0050
EM8.20753.0050CS
29587.63
USD
EM8.20753.0050
CA8.20753.0050
Lithium hydride for synthesis, Sigma-Aldrich®
Lithium hydride
Formula:
HLi |
MDL Number:
MFCD00011074 CAS Number: 7580-67-8 UN: 1414 ADR: 4.3,I |
Specification Test Results
Solubility | (20 °C) (rigorous decomposition) |
Melting Point | 688 °C |
Molar Mass | 7.95 g/mol |
Bulk Density | 290 - 430 kg/m3 |
Density | 0.78 g/cm3 (20 °C) |
pH |
|
Hazard Class | 4.3 |
Hazard Class Description | Dangerous When Wet Material |
Hazard Class Description | Dangerous When Wet Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...