Potassium tetrahydroborate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.20747.0025
8.20747.0100
8.20747.1000
8.20747.1000
8.20747.0100
8.20747.0025
EM8.20747.1000CS
2239
USD
EM8.20747.1000
CA8.20747.1000
CA8.20747.0025
CA8.20747.0100
EM8.20747.0025
EM8.20747.0100
Potassium tetrahydroborate for synthesis, Sigma-Aldrich®
Potassium tetrahydroborate
Formula:
H₄BK MW: 53.94 g/mol |
MDL Number:
MFCD00011396 CAS Number: 13762-51-1 UN: 1870 ADR: 4.3,I |
Specification Test Results
Solubility | (20 °C) (rigorous decomposition) |
Melting Point | 500 °C |
Molar Mass | 53.94 g/mol |
Bulk Density | 400 kg/m3 |
Density | 1.11 g/cm3 |
Hazard Class | 4.3 |
Hazard Class Description | Dangerous When Wet Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...