Phenyl glycidyl ether for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Glycidyl phenyl ether
M-Clarity™ quality level = MQ200
8.20553.0250
8.20553.0250
EM8.20553.0250CS
793
USD
EM8.20553.0250
CA8.20553.0250
Phenyl glycidyl ether for synthesis, Sigma-Aldrich®
Phenyl glycidyl ether
Formula:
C₉H₁₀O₂ MW: 150.18 |
MDL Number:
MFCD00005133 CAS Number: 122-60-1 |
Specification Test Results
Solubility | 2.4 g/l (20 °C) |
Melting Point | 3.5 °C |
Molar Mass | 150.17 g/mol |
Boiling Point | 244 - 246 °C |
Vapor Pressure | 0.013 hPa (25 °C) |
Flash Point | 114 °C |
Refractive Index | 1.5305 (20 °C, 589 nm) |
Density | 1.11 g/cm3 (20 °C) |
Ignition Point | 430 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...