Borane dimethylamine complex for synthesis, Sigma-Aldrich®
|
Danger
|
Synonyms:
Boron hydride dimethylamine
, Borane, dimethylamine complex
M-Clarity™ quality level = MQ200
8.20496.0050
8.20496.0010
8.20496.0050
EM8.20496.0050CS
3003.74
USD
EM8.20496.0050
CA8.20496.0010
CA8.20496.0050
Borane dimethylamine complex for synthesis, Sigma-Aldrich®
Borane dimethylamine complex
Formula:
C₂H₁₀BN MW: 58.92 g/mol |
UN:
2926 ADR: 4.1 (6.1),II |
Specification Test Results
Solubility | 128 g/l (20 °C) |
Melting Point | 33 - 36 °C |
Molar Mass | 58.92 g/mol |
Bulk Density | 300 - 350 kg/m3 |
Vapor Pressure |
|
Flash Point | 65 °C |
Density | 0.73 g/cm3 (20 °C) |
pH | 8 - 9 (10 g/l, H2O, 20 °C) |
Ignition Point | 175 °C |
Hazard Class | 4.1 |
Hazard Class Description | Flammable Solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...