Luminol (3-Aminophthalhydrazide) for synthesis, Sigma-Aldrich®
Synonyms:
5-Amino-2,3-dihydrophthalazine-1,4-dione
, NSC 5064
, 5-Amino-2,3-Dihydro-1,4-Phthalazinedione
M-Clarity™ quality level = MQ200
8.20071.0001
EM8.20071.0001EA
61
USD
EM8.20071.0001
Luminol (3-Aminophthalhydrazide) for synthesis, Sigma-Aldrich®
Luminol (3-Aminophthalhydrazide)
Formula:
C₈H₇N₃O₂ MW: 177.16 g/mol |
MDL Number:
MFCD00006890 CAS Number: 521-31-3 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 318 - 320 °C |
Molar Mass | 177.16 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...