Lithium aluminum hydride, tablets for synthesis, Sigma-Aldrich®

Danger

Synonyms: Lithium tetrahydridoaluminateAluminium lithium hydride

M-Clarity™ quality level = MQ200

8.18878.0250 8.18878.0250
EM8.18878.0250CS 12982.08 USD
EM8.18878.0250 CA8.18878.0250
Lithium aluminum hydride, tablets for synthesis, Sigma-Aldrich®
Lithium aluminum hydride
Formula: H₄AlLi
MW: 37.95 g/mol
MDL Number: MFCD00011075
CAS Number: 16853-85-3
UN: 1410
ADR: 4.3,I

Order Now

Specification Test Results

Solubility (20 °C) (rigorous decomposition)
Molar Mass 37.95 g/mol
Bulk Density 400 kg/m3
Density 0.917 g/cm3 (20 °C)
Hazard Class 4.3
Hazard Class Description Dangerous When Wet Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR