Bromoform ≥98.0% stabilized for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Tribromomethane
Stabilization:
Stabilised with 2-methyl-2-butene (iso-amylene)
M-Clarity™ quality level = MQ200
8.18702.0250
8.18702.1000
8.18702.1000
8.18702.0250
EM8.18702.1000CS
2905
USD
EM8.18702.1000
CA8.18702.0250
CA8.18702.1000
EM8.18702.0250
Bromoform ≥98.0% stabilized for synthesis, Sigma-Aldrich®
Bromoform
Formula:
Br₃CH MW: 252.73 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00000128 CAS Number: 75-25-2 UN: 2515 ADR: 6.1,III |
Specification Test Results
Solubility | 1 g/l (20 °C) |
Melting Point | 9.2 °C |
Molar Mass | 252.73 g/mol |
Boiling Point | 149.5 °C (1013 mbar) |
Vapor Pressure | 6.4 hPa (20 °C) |
Density | 2.89 g/cm3 (20 °C) |
Hazard Class | 6.1 |
Hazard Class Description | Toxic Materials |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...