Diethyl isobutylidenemalonate for synthesis, Sigma-Aldrich®
M-Clarity™ quality level = MQ200
8.18389.0050
8.18389.0050
EM8.18389.0050CS
6093.11
USD
CA8.18389.0050
EM8.18389.0050
Diethyl isobutylidenemalonate for synthesis, Sigma-Aldrich®
Diethyl isobutylidenemalonate
Specification Test Results
Solubility | (20 °C) insoluble |
Molar Mass | 214.26 g/mol |
Boiling Point | 117 - 119 °C (17 hPa) |
Density | 0.99 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...