(2S,5SR)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropyl-5-methylpyrazine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.18316.0001 8.18316.0001
EM8.18316.0001CS 13052 USD
EM8.18316.0001 CA8.18316.0001
(2S,5SR)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropyl-5-methylpyrazine for synthesis, Sigma-Aldrich®
(2S,5SR)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropyl-5-methylpyrazine
Formula: C₁₀H₁₈N₂O₂
CAS Number: 116907-51-8

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 198.26 g/mol
Boiling Point 103 - 104 °C (21 hPa)
Density 1.00 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR