Poly(methyl hydrogen siloxane) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.18063.1000 8.18063.0250 8.18063.1000 8.18063.0250
EM8.18063.0250CS 1292 USD
CA8.18063.1000 CA8.18063.0250 EM8.18063.0250 EM8.18063.1000
Poly(methyl hydrogen siloxane) for synthesis, Sigma-Aldrich®
Poly(methyl hydrogen siloxane)
Formula: C₇H₂₁O₂Si₃
CAS Number: 26403-67-8

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Vapor Pressure 25 hPa (20 °C)
Flash Point 100 °C
Refractive Index 1.395 (20 °C, 589 nm)
Density 1.00 g/cm3 (20 °C)
pH 7 (H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR