Sodium cyanotrihydroborate for synthesis

Supplier: MilliporeSigma

Synonyms: Sodium cyanoborohydride

8.18053.0005 8.18053.0005 8.18053.0250 8.18053.0250
EM8.18053.0005CS 3384 USD InStock
CA8.18053.0005 EM8.18053.0005 EM8.18053.0250 CA8.18053.0250
Sodium cyanotrihydroborate for synthesis
Sodium cyanotrihydroborate
Formula: CH₃BNNa
MW: 62,84 g/mol
Melting Pt: 241 to 243 °C
Density: 1.11 to 1.12 g/cm³ (28 °C)
Storage Temperature: ambient
MDL Number: MFCD00003516
CAS Number: 25895-60-7
EINECS: 247-317-2
UN: 3134
ADR: 4.3,II
Merck Index: 13,08680

Order Now

Specification Test Results

Solubility 2100 g/l (20 °C) (slow decomposition)
Melting Point 241 - 243 °C
Molar Mass 62.84 g/mol
Hazard Class 4.3
Hazard Class Description Dangerous When Wet Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR