2,4-Diquinolinecarboxylic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Quinoline-2,4-dicarboxylic acid
M-Clarity™ quality level = MQ200
8.15103.0025
8.15103.0025
EM8.15103.0025CS
18296
USD
EM8.15103.0025
CA8.15103.0025
2,4-Diquinolinecarboxylic acid for synthesis, Sigma-Aldrich®
2,4-Diquinolinecarboxylic acid
Specification Test Results
Melting Point | 229 °C |
Molar Mass | 217.18 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...