Ethylene glycol-tert-butyl ethyl ether for synthesis, Sigma-Aldrich®
|
Warning
|
M-Clarity™ quality level = MQ200
8.14985.0100
8.14985.0100
EM8.14985.0100CS
310.5
USD
EM8.14985.0100
CA8.14985.0100
Ethylene glycol-tert-butyl ethyl ether for synthesis, Sigma-Aldrich®
Ethylene glycol tert-butyl ethyl ether
Formula:
C₈H₁₈O₂ |
UN:
1993 ADR: 3,III |
Specification Test Results
Solubility | 30 g/l |
Melting Point | -70 °C |
Molar Mass | 146.23 g/mol |
Boiling Point | 155 °C (1013 hPa) |
Vapor Pressure | 4 hPa |
Flash Point | 27 °C |
Explosion Limit | 0.8 - 8.5 %(V) |
Density | 0.83 g/cm3 (20 °C) |
pH | 7 (H2O) |
Ignition Point | 230 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...