1-Bromo-4-(trifluoromethoxy)benzene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
4-(Trifluoromethoxy)bromobenzene
M-Clarity™ quality level = MQ200
8.14964.0050
8.14964.0050
EM8.14964.0050CS
15429
USD
CA8.14964.0050
EM8.14964.0050
1-Bromo-4-(trifluoromethoxy)benzene for synthesis, Sigma-Aldrich®
1-Bromo-4-(trifluoromethoxy)benzene
Formula:
C₇H₄BrF₃O MW: 241.01 g/mol |
MDL Number:
MFCD00040834 CAS Number: 407-14-7 ADR: 9, |
Specification Test Results
Solubility | 11.7 mg/l (20 °C) |
Molar Mass | 241.01 g/mol |
Boiling Point | 91 - 92 °C (100 hPa) |
Vapor Pressure | 20 hPa (55 °C) |
Flash Point | 67 °C |
Density | 1.62 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...