Ethyl-2-aminobenzo[b]thiophene-3-carboxylate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Ethyl 2-aminobenzo[b]thiophene-3-carboxylate
M-Clarity™ quality level = MQ200
8.14873.0005
8.14873.0005
EM8.14873.0005CS
229069.9
USD
EM8.14873.0005
CA8.14873.0005
Ethyl-2-aminobenzo[b]thiophene-3-carboxylate for synthesis, Sigma-Aldrich®
Ethyl 2-aminobenzo[b]thiophene-3-carboxylate
Formula:
C₁₁H₁₁NO₂S MW: 221.2755 g/mol |
MDL Number:
MFCD00102506 CAS Number: 7311-95-7 UN: 2811 ADR: 6.1,II |
Specification Test Results
Melting Point | 108.6 °C |
Molar Mass | 221.28 g/mol |
Boiling Point |
|
Vapor Pressure | (20 °C) |
Hazard Class | 6.1 |
Hazard Class Description | Toxic Materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...