N,N-Diethylhydroxylamine ≥85% in water for synthesis, Sigma-Aldrich®
|
Warning
|
M-Clarity™ quality level = MQ200
8.14671.0250
8.14671.0250
8.14671.1000
8.14671.1000
EM8.14671.0250CS
698.02
USD
EM8.14671.0250
EM8.14671.1000
CA8.14671.1000
CA8.14671.0250
N,N-Diethylhydroxylamine ≥85% in water for synthesis, Sigma-Aldrich®
N,N-Diethylhydroxylamine
Formula:
C₄H₁₁NO |
MDL Number:
MFCD00002126 CAS Number: 3710-84-7 UN: 1993 ADR: 3,III |
Specification Test Results
Solubility | (20 °C) soluble |
Boiling Point | 125 - 130 °C (1013 hPa) |
Vapor Pressure | 43 hPa (25 °C) |
Flash Point | 46 °C |
Density | 0.9 g/cm3 (20 °C) |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...