3-Methoxycyclohex-2-en-1-one for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.14561.0010
8.14561.0010
EM8.14561.0010CS
133217
USD
CA8.14561.0010
EM8.14561.0010
3-Methoxycyclohex-2-en-1-one for synthesis, Sigma-Aldrich®
3-Methoxycyclohex-2-en-1-one
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 32 - 35 °C |
Molar Mass | 126.16 g/mol |
Boiling Point | 114 °C (19 hPa) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...