(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
(S)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide
M-Clarity™ quality level = MQ200
8.14473.0001
8.14473.0001
EM8.14473.0001CS
249357
USD
EM8.14473.0001
CA8.14473.0001
(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone for synthesis, Sigma-Aldrich®
(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone
Formula:
C₂₅H₂₄N₂O₂ MW: 384.470 g/mol |
MDL Number:
MFCD00145260 CAS Number: 96293-17-3 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 97 - 99 °C |
Molar Mass | 384.47 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...