4,5-Difluorophthalic anhydride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
4,5-Difluorophthalic Anhydride
M-Clarity™ quality level = MQ200
8.14389.0001
8.14389.0001
EM8.14389.0001CS
337104
USD
CA8.14389.0001
EM8.14389.0001
4,5-Difluorophthalic anhydride for synthesis, Sigma-Aldrich®
4,5-Difluorophthalic anhydride
Formula:
C₈H₂F₂O₃ MW: 184.1 g/mol |
MDL Number:
MFCD08695033 UN: 3261 ADR: 8,III |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 92 - 96 °C |
Molar Mass | 184.09 g/mol |
Boiling Point | 92 - 96 °C (14 hPa) |
Flash Point | 73 °C |
Density | 0.975 g/cm3 |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...