Glycolic acid ≥70% in water for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
2-Hydroxy acid
, 2-Hydroxyacetic acid
M-Clarity™ quality level = MQ200
8.14186.1000
8.14186.1000
EM8.14186.1000CS
539
USD
CA8.14186.1000
EM8.14186.1000
Glycolic acid ≥70% in water for synthesis, Sigma-Aldrich®
Glycolic acid
Formula:
C₂H₄O₃ MW: 76.05 g/mol |
MDL Number:
MFCD00004312 CAS Number: 79-14-1 UN: 3265 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 10 °C |
Boiling Point | 112 °C (1013 hPa) |
Density | 1.262 g/cm3 (20 °C) |
pH | 0.1 (H2O, 25 °C) |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...