Disulfur dichloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Sulfur monochloride
, Sulphur monochloride
M-Clarity™ quality level = MQ200
8.14179.1000
8.14179.1000
8.14179.0100
EM8.14179.0100CS
491
USD
EM8.14179.0100
EM8.14179.1000
CA8.14179.1000
Disulfur dichloride for synthesis, Sigma-Aldrich®
Disulphur dichloride
Formula:
Cl₂S₂ MW: 135.04 g/mol |
CAS Number:
10025-67-9 UN: 1828 ADR: 8,I |
Specification Test Results
Solubility | (20 °C) (rigorous decomposition) |
Melting Point | -76 °C |
Molar Mass | 135.04 g/mol |
Boiling Point | 138 °C |
Vapor Pressure | 14.7 hPa (20 °C) |
Flash Point | 4.2 - 32.5 %(V) |
Refractive Index |
|
Explosion Limit | 1.67 (20 °C, 589 nm) |
Density | 1.68 g/cm3 (20 °C) |
Ignition Point | 390 °C |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...