Polyethylene glycol dimethylether, M.W. 250 for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.14173.0500
8.14173.0500
EM8.14173.0500CS
946
USD
EM8.14173.0500
CA8.14173.0500
Polyethylene glycol dimethylether, M.W. 250 for synthesis, Sigma-Aldrich®
Polyethylene glycol dimethylether 150
Formula:
H₃C(OCH₂CH₂)nOCH₃ |
MDL Number:
MFCD00147972 CAS Number: 24991-55-7 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -23 °C |
Molar Mass | 250 g/mol |
Boiling Point |
|
Vapor Pressure |
|
Flash Point | 137 °C |
Refractive Index | 1.439 (20 °C, 589 nm) |
Explosion Limit | 0.5 %(V) |
Density | 1.03 g/cm3 (20 °C) |
pH | (100 g/l, H2O, 20 °C) neutral |
Viscosity kinematic | 7.8 mm2/s (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...