Ethylene glycolbismercaptoacetate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Ethylene Mercaptoacetate
, Ethylene Bis(thioglycolate)
M-Clarity™ quality level = MQ200
8.14145.0100
8.14145.0100
EM8.14145.0100CS
1859
USD
CA8.14145.0100
EM8.14145.0100
Ethylene glycolbismercaptoacetate for synthesis, Sigma-Aldrich®
Ethylene glycol bismercaptoacetate
Formula:
C₆H₁₀O₄S₂ MW: 210.28 g/mol |
MDL Number:
MFCD00004875 UN: 3082 ADR: 9,III |
Specification Test Results
Solubility | 20 g/l (20 °C) |
Molar Mass | 210.27 g/mol |
Boiling Point | 137 - 139 °C (2 hPa) |
Density | 1.32 g/cm3 (20 °C) |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous Hazardous Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...