Valeraldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: AmylaldehydePentanalPentylaldehyde

M-Clarity™ quality level = MQ200

8.08504.0100 8.08504.0100 8.08504.0500 8.08504.0500
EM8.08504.0500CS 397 USD
CA8.08504.0500 CA8.08504.0100 EM8.08504.0500 EM8.08504.0100
Valeraldehyde for synthesis, Sigma-Aldrich®
Valeraldehyde
Formula: C₅H₁₀O
MW: 86.13 g/mol
MDL Number: MFCD00007026
CAS Number: 110-62-3
UN: 2058
ADR: 3,II

Order Now

Specification Test Results

Solubility 14 g/l (20 °C)
Melting Point -92 °C
Molar Mass 86.13 g/mol
Boiling Point 102 °C (1013 hPa)
Vapor Pressure 36 hPa (20 °C)
Flash Point 7 °C
Refractive Index 1.3942 (20 °C, 589 nm)
Explosion Limit 1.7 - 6.8 %(V)
Density 0.81 g/cm3 (20 °C)
pH (H2O) neutral
Ignition Point 205 °C
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR