Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Pentylamine for synthesis

Supplier: MilliporeSigma

Synonyms: Amylaminen-Amylamine1-Aminopentanen-Pentylamine1-Pentylamine

8.07333.0025 8.07333.0100 8.07333.0025 8.07333.0100
EM8.07333.0100CS 3208.06 USD InStock
CA8.07333.0025 EM8.07333.0100 EM8.07333.0025 CA8.07333.0100
Pentylamine for synthesis
Pentylamine


Formula: H₃C(CH₂)₃CH₂NH₂
MW: 87.17 g/mol
Boiling Pt: 104.4 °C (1013 hPa)
Melting Pt: -50 °C
Density: 0.7547 g/cm³ (20 °C)
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00008236
CAS Number: 110-58-7
EINECS: 203-780-2
UN: 1106
ADR: 3,II
Merck Index: 13,00602
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -50 °C
Molar Mass 87.16 g/mol
Boiling Point 103 °C (1013 hPa)
Vapor Pressure 120.9 hPa (47 °C)
Flash Point 7 °C
Refractive Index 1.411 (20 °C, 589 nm)
Explosion Limit 2.2 - 22 %(V)
Density 0.75 g/cm3 (20 °C)
Ignition Point 305 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty