Lithium aluminum hydride, powder for synthesis, Sigma-Aldrich®
|
Danger
|
Synonyms:
Lithium tetrahydridoaluminate
, Aluminium lithium hydride
M-Clarity™ quality level = MQ200
8.05661.0010
CA8.05661.0010
Lithium aluminum hydride, powder for synthesis, Sigma-Aldrich®
Lithium aluminum hydride
Formula:
H₄AlLi MW: 37.95 g/mol Melting Pt: 125 °C Density: 0.91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011075 CAS Number: 16853-85-3 EINECS: 240-877-9 UN: 1410 ADR: 4.3,I Merck Index: 13,00348 |
Specification Test Results
Solubility | (20 °C) (rigorous decomposition) |
Melting Point | 125 °C (decomposition) |
Molar Mass | 37.95 g/mol |
Bulk Density | 400 kg/m3 |
Density | 0.917 g/cm3 (20 °C) |
pH | (H2O) alkaline |
Hazard Class | 4.3 |
Hazard Class Description | Dangerous When Wet Material |
Hazard Class Description | Dangerous When Wet Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...