Furfurylamine for synthesis, Sigma-Aldrich®
|
Danger
|
Synonyms:
2-(Aminomethyl)furan
M-Clarity™ quality level = MQ200
8.04016.0250
8.04016.0250
EM8.04016.0250CS
2343.53
USD
CA8.04016.0250
EM8.04016.0250
Furfurylamine for synthesis, Sigma-Aldrich®
Furfurylamine
Formula:
C₅H₇NO MW: 97.12 g/mol |
MDL Number:
MFCD00003258 CAS Number: 617-89-0 EINECS: 210-536-9 UN: 2526 ADR: 3 (8),III |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -70 °C |
Molar Mass | 97.11 g/mol |
Boiling Point | 145 - 146 °C (1013 hPa) |
Vapor Pressure | 4 hPa (20 °C) |
Flash Point | 46 °C DIN 51755 Part 1 |
Refractive Index | 1.49 (20 °C, 589 nm) |
Explosion Limit | 1.8 %(V) |
Density | 1.05 g/cm3 (20 °C) |
pH | 11.6 (100 g/l, H2O, 20 °C) |
Ignition Point | 350 °C DIN 51794 |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...