Cumene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-PhenylpropaneIsopropylbenzene

M-Clarity™ quality level = MQ200

8.02681.0100 8.02681.0100
EM8.02681.0100CS 566 USD
EM8.02681.0100 CA8.02681.0100
Cumene for synthesis, Sigma-Aldrich®
Cumene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: –96 °C
Density: 0.8624 g/cm³ (20 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00008881
CAS Number: 98-82-8
EINECS: 202-704-5
UN: 1918
ADR: 3,III
Merck Index: 13,02644

Order Now

Specification Test Results

Solubility 0.05 g/l (20 °C)
Melting Point -96 °C
Molar Mass 120.19 g/mol
Boiling Point 153 °C (1013 hPa)
Vapor Pressure 5.3 hPa (20 °C)
Flash Point 0.8 - 6.0 %(V)
Refractive Index 31 °C
Density 0.86 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 420 °C
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR