Benzophenone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: DiphenylmethanoneDiphenyl ketone

M-Clarity™ quality level = MQ200

8.01801.0005 8.01801.0005
EM8.01801.0005CS 15525 USD
CA8.01801.0005 EM8.01801.0005
Benzophenone for synthesis, Sigma-Aldrich®
Benzophenone
Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 305.4 °C (1013 hPa)
Melting Pt: 47…49 °C
Density: 1.085 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
UN: 3077
ADR: 9,III
Merck Index: 13,01099

Order Now

Specification Test Results

Solubility 0.14 g/l (25 °C)
Melting Point 47 - 49 °C
Molar Mass 182.22 g/mol
Bulk Density 700 kg/m3
Boiling Point 304 - 306 °C (1013 hPa)
Vapor Pressure 0.004 hPa (20 °C)
Flash Point 150 °C
Density 1.1 g/cm3 (25 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR