2-Bromobutane for synthesis, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ200

8.01548.1000 8.01548.1000
EM8.01548.1000CS 1558.73 USD
EM8.01548.1000 CA8.01548.1000
2-Bromobutane for synthesis, Sigma-Aldrich®
2-Bromobutane
Formula: H₃CBrCHCH₂CH₃
MW: 137.02 g/mol
Boiling Pt: 91 °C (1013 hPa)
Melting Pt: –112 °C
Density: 1.258 g/cm³ (20 °C)
Flash Pt: 21 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000156
CAS Number: 78-76-2
EINECS: 201-140-7
UN: 2339
ADR: 3,II
Merck Index: 13,01553

Order Now

Specification Test Results

Solubility  - 1 g/l (20 °C)
Melting Point -112 °C
Molar Mass 137.01 g/mol
Boiling Point 90 - 92 °C (1013 hPa)
Vapor Pressure 70 hPa (20 °C)
Flash Point 21 °C
Refractive Index 1.4369 (20 °C, 589 nm)
Explosion Limit 2.6 - 6.6 %(V)
Density 1.26 g/cm3 (20 °C)
Ignition Point 265 °C
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR