Potassium permanganate 0.05 mol/l (0.25 N) in aqueous solution, Titripur® volumetric solution, Supelco®


M-Clarity™ quality level = MQ100

4.80160.2500
CA4.80160.2500
Potassium permanganate 0.05 mol/l (0.25 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

pH 4 - 8 (H2O)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR