Extran® AP 33 Liquid Anti-Foaming Agent for Automatic Washing, MilliporeSigma
Supplier: MilliporeSigma
Extran®
1.40007.2500
CA1.40007.2500
Extran® AP 33 Liquid Anti-Foaming Agent for Automatic Washing, MilliporeSigma
Cleaning Agents Detergents and Surface Cleansers
When diluted with water, concentrates can clean by themselves without leaving residues
For routine laboratory applications, no scrubbing is necessary once items are removed from the cleaning bath. For best results, items to be cleaned should be soaked for 2 to 24 hours, and rinsed with distilled water immediately after removal from cleaning bath.