Standard oil for atomic absorption spectroscopy, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

1.13898.2500 1.13898.2500
EM1.13898.2500CS 2203 USD
CA1.13898.2500 EM1.13898.2500
Standard oil for atomic absorption spectroscopy, Supelco®
Standard oil

Standard oil, solvent for atomic absorption spectroscopy

Density: 0.85 g/cm3 (20 °C)
Flash Pt: >180 °C
Storage Temperature: Ambient
CAS Number: 8042-47-5

Order Now

Specification Test Results

Identity (IR-spectrum) passes test
Density (d 20 °C/ 4 °C) 0.84 - 0.86 g/ml
Viscosity (20 °C) 30 - 40 mPa∙s
Ag (Silver) ≤ 0.5 ppm
Al (Aluminium) ≤ 0.5 ppm
Au (Gold) ≤ 0.5 ppm
B (Boron) ≤ 0.5 ppm
Ba (Barium) ≤ 0.5 ppm
Be (Beryllium) ≤ 0.5 ppm
Bi (Bismuth) ≤ 0.5 ppm
Ca (Calcium) ≤ 0.5 ppm
Cd (Cadmium) ≤ 0.5 ppm
Co (Cobalt) ≤ 0.5 ppm
Cr (Chromium) ≤ 0.5 ppm
Cu (Copper) ≤ 0.5 ppm
Fe (Iron) ≤ 0.5 ppm
Ga (Gallium) ≤ 0.5 ppm
In (Indium) ≤ 0.5 ppm
K (Potassium) ≤ 0.5 ppm
Li (Lithium) ≤ 0.5 ppm
Mg (Magnesium) ≤ 0.5 ppm
Mn (Manganese) ≤ 0.5 ppm
Mo (Molybdenum) ≤ 0.5 ppm
Na (Sodium) ≤ 0.5 ppm
Ni (Nickel) ≤ 0.5 ppm
Pb (Lead) ≤ 0.5 ppm
Pt (Platinum) ≤ 0.5 ppm
Sb (Antimony) ≤ 0.5 ppm
Sn (Tin) ≤ 0.5 ppm
Sr (Strontium) ≤ 0.5 ppm
Ti (Titanium) ≤ 0.5 ppm
Tl (Thallium) ≤ 0.5 ppm
V (Vanadium) ≤ 0.5 ppm
Zn (Zinc) ≤ 0.5 ppm
Zr (Zirconium) ≤ 0.5 ppm
Water ≤ 0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR