Bismuth(III) chloride, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Bismuth trichloride
M-Clarity™ quality level = MQ200
1.12403.0100
1.12403.0100
EM1.12403.0100CS
16824
USD
CA1.12403.0100
EM1.12403.0100
Bismuth(III) chloride, Sigma-Aldrich®
Bismuth (III) chloride
Formula:
BiCl₃ MW: 315.34 g/mol Boiling Pt: 447 °C (1013 hPa) Melting Pt: 232 °C Density: 4.75 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003461 CAS Number: 7787-60-2 EINECS: 232-123-2 ADR: 8,III Merck Index: 13,01261 |
Specification Test Results
Solubility | (20 °C) soluble,Hydrolysis |
Melting Point | 230 - 232 °C |
Molar Mass | 315.33 g/mol |
Boiling Point | 447 °C (1013 hPa) |
Density | 4.75 g/cm3 (20 °C) |
pH | (H2O, 20 °C) strongly acid,Hydrolysis |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...