Silver nitrate 0.1 M concentrated aqueous solution, Titrisol®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

1.09990.0001 1.09990.0001
EM1.09990.0001CS 8650 USD
EM1.09990.0001 CA1.09990.0001
Silver nitrate 0.1 M concentrated aqueous solution, Titrisol®, Supelco®
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1760
ADR: 8,
https://us.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.27 g/cm3 (20 °C)
pH 7 - 9 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR