Sodium hydroxide 0.01 M concentrated aqueous solution, Titrisol®, Supelco®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
1.09961.0001
1.09961.0001
EM1.09961.0001CS
1887
USD
EM1.09961.0001
CA1.09961.0001
Sodium hydroxide 0.01 M concentrated aqueous solution, Titrisol®, Supelco®
Sodium hydroxide
Formula:
NaOH MW: 40 g/mol Boiling Pt: ∼105 °C (1013 hPa) Density: 1.327 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8, |
Specification Test Results
Solubility | (20 °C) soluble |
Density | 1.01 g/cm3 (20 °C) |
pH | 13 (H2O, 20 °C) |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...