Sodium hydroxide 1 N concentrated aqueous solution, Titrisol®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

M-Clarity™ quality level = MQ100

1.09956.0001 1.09956.0001
EM1.09956.0001CS 1731 USD
CA1.09956.0001 EM1.09956.0001
Sodium hydroxide 1 N concentrated aqueous solution, Titrisol®, Supelco®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: ∼105 °C (1013 hPa)
Density: 1.327 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.030 g/cm3 (20 °C)
pH 14 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR