Potassium permanganate 0,002 mol/l (0.01 N) concentrated aqueous solution, Titrisol®, Supelco®
|
Warning
|
M-Clarity™ quality level = MQ100
1.09930.0001
CA1.09930.0001
Potassium permanganate 0,002 mol/l (0.01 N) concentrated aqueous solution, Titrisol®, Supelco®
Potassium permanganate
Formula:
KMnO₄ MW: 158.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011364 CAS Number: 7722-64-7 EINECS: 231-760-3 UN: 3082 ADR: 9,III |
Specification Test Results
Solubility | (20 °C) soluble |
Density | 1.002 g/cm3 (20 °C) |
pH | 7.3 (H2O, 20 °C) |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous Hazardous Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...