2',7'-Dichlorofluorescein indicator, Supelco®
Supplier: MilliporeSigma
Synonyms:
2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one
1.09676.0005
1.09676.0005
EM1.09676.0005CS
88513
USD
CA1.09676.0005
EM1.09676.0005
2',7'-Dichlorofluorescein indicator, Supelco®
2',7'-Dichlorofluorescein
Formula:
C₂₀H₁₀Cl₂O₅ MW: 401.20 g/mol |
MDL Number:
MFCD00005047 CAS Number: 76-54-0 EINECS: 200-968-6 |
Specification Test Results
Solubility | (20 °C) insoluble |
Molar Mass | 401.2 g/mol |
Bulk Density | 390 kg/m3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...