Alkali blue (C.I. 42765) indicator, Supelco®

Supplier: MilliporeSigma

1.09196.0025 1.09196.0025
EM1.09196.0025CS 1881 USD
CA1.09196.0025 EM1.09196.0025
Alkali blue (C.I. 42765) indicator, Supelco®
Alkali blue (C.I. 42765)
Formula: C₃₇H₂₉N₃O₃S
MW: 595.73 g/mol
MDL Number: MFCD00083221
CAS Number: 1324-76-1

Order Now

Specification Test Results

Molar Mass 595.73 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR