Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution, Titripur® Reag. USP volumetric solution, Supelco®

Warning


M-Clarity™ quality level = MQ100

1.09122.1000
EM1.09122.1000CS 811.81 USD
EM1.09122.1000
Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution, Titripur® Reag. USP volumetric solution, Supelco®
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.01 g/cm3 (20 °C)
pH 8 (H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR