Trifluoromethanesulfonic acid 0.1 mol/l in anhydrous acetic acid, Titripur®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Trifluoromethanesulfonic acidTrifluoromethanesulphonic acid

1.08450.1000 1.08450.1000
EM1.08450.1000CS 958 USD
CA1.08450.1000 EM1.08450.1000
Trifluoromethanesulfonic acid 0.1 mol/l in anhydrous acetic acid, Titripur®, Supelco®
Trifluoromethanesulphonic acid
Formula: CHF₃O₃S
MW: 150.08 g/mol
Boiling Pt: 118 °C (1013 hPa)
Density: 1.06 g/cm3 (20 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00007514
CAS Number: 1493-13-6
UN: 2920
ADR: 8 (3),II

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C. \< - 0.5 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR