Strontium nitrate, anhydrous, Suprapur® for trace metal analysis, Supelco®
|
Danger
|
1.07871.0250
1.07871.0050
1.07871.0250
1.07871.0050
EM1.07871.0250CS
19952.01
USD
EM1.07871.0250
EM1.07871.0050
CA1.07871.0250
CA1.07871.0050
Strontium nitrate, anhydrous, Suprapur® for trace metal analysis, Supelco®
Strontium nitrate
Formula:
Sr(NO₃)₂ MW: 211.63 g/mol Boiling Pt: 645 °C (1013 hPa) Melting Pt: 570 °C Density: 2.113 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011248 CAS Number: 10042-76-9 EINECS: 233-131-9 UN: 1507 ADR: 5.1,III Merck Index: 12,09007 |
Specification Test Results
Solubility | 660 g/l (20 °C) |
Melting Point | 570 °C |
Molar Mass | 211.63 g/mol |
Bulk Density | 1000 kg/m3 |
Boiling Point | 645 °C |
Density | 2.986 g/cm3 (20 °C) |
pH | 5 - 7 (50 g/l, H2O, 20 °C) |
Hazard Class | 5.1 |
Hazard Class Description | Oxidizer |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...