Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silica gel 60 HF₂₅₄+₃₆₆ for thin-layer chromatography

Supplier: MilliporeSigma
1.07741.1000
Standardized sorbents ensure reliable results every time. EMD Millipore's broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µ: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Size pH Molar Mass Packaging Weight H x L x W Melting Point Density Type Supplier No. VWR Catalog Number Unit Price Quantity
Silica gel 60 HF₂₅₄+₃₆₆ for thin-layer chromatography 1 kg 7 (100 g/l, H2O, 20 °C) (slurry) 60.08 g/mol Plastic Bottle 0.6096 lbs. 22 x 18 x 18" 1710° C 200 - 800 kg/m3 (Bulk) Chemical 1.07741.1000 EM1.07741.1000 Each (1kg) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Solubility (20° C) insoluble
Chemical Formula Silica gel
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty